Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mkl1 Antibody (OTI3G7), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272715B
Description
Mkl1 Monoclonal specifically detects Mkl1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Mkl1 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
MRTFA | |
Recombinant protein fragment corresponding to amino acids 165-397 of human Mkl1 (NP_065882) produced in E.coli. | |
0.1 mL | |
57591.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3G7 | |
Western Blot, Immunohistochemistry | |
AMKL, KIAA1438, MALSAP and coiled-coil domain, megakaryoblastic leukemia (translocation) 1, Megakaryoblastic leukemia 1 protein, Megakaryocytic acute leukemia protein, MKL/myocardin-like protein 1, Myocardin-related transcription factor A, RNA-binding motif protein 15/megakaryoblastic leukemia-1 fusion protein | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction