Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MKL1 Monoclonal Antibody (CL4281)
GREENER_CHOICE

Catalog No. PIMA531399
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531399 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531399 Supplier Invitrogen™ Supplier No. MA531399
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: EQEKRAQQPA PAPAPLGTPV KQENSFSSCQ LSQQPLGPAH PFNPSLAAPA TNHIDPCAVA PGPPSVVVKQ EALQPEPEPV PAPQLLLGPQ GPSLIKGVAP PTLITDSTGT HLVLTVTNKN ADSPG Highest antigen sequence indentity to the following orthologs: Rat - 66%, Mouse - 65%.

The protein encoded by this gene interacts with the transcription factor myocardin, a key regulator of smooth muscle cell differentiation. The encoded protein is predominantly nuclear and may help transduce signals from the cytoskeleton to the nucleus. This gene is involved in a specific translocation event that creates a fusion of this gene and the RNA-binding motif protein-15 gene. This translocation has been associated with acute megakaryocytic leukemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MKL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL4281
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Mrtfa
Gene Accession No. Q969V6
Gene Alias AMKL; basic SAP and coiled-coil domains; basic SAP coiled-coil transcription activator; basic, SAP and coiled-coil domain; basic, SAP, and coiled-coil domain containing protein; Bsac; FLMKL1; KIAA1438; Mal; megakaryoblastic leukemia (translocation) 1; megakaryoblastic leukemia (translocation) 1 homolog; megakaryoblastic leukemia 1 met; megakaryoblastic leukemia 1 protein; Megakaryoblastic leukemia 1 protein homolog; Megakaryocytic acute leukemia protein; Megakaryocytic acute leukemia protein homolog; MELODY; mixed lineage leukemia; MKL (megakaryoblastic leukemia)/myocardin-like 1; MKL/myocardin-like protein 1; MKL1; MKL1-elongated derivative of yield protein; MKL1met; MRTFA; MRTF-A; myocardin related transcription factor A; myocardin-related transcription factor A
Gene Symbols Mrtfa
Host Species Mouse
Immunogen Recombinant protein fragment of Human MKL1 (Product #RP-91546)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57591
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.