Learn More
Invitrogen™ MKL1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total MKL1 protein.
Specifications
Specifications
| Antigen | MKL1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Mrtfa |
| Gene Accession No. | Q969V6 |
| Gene Alias | AMKL; basic SAP and coiled-coil domains; basic SAP coiled-coil transcription activator; basic, SAP and coiled-coil domain; basic, SAP, and coiled-coil domain containing protein; Bsac; FLMKL1; KIAA1438; Mal; megakaryoblastic leukemia (translocation) 1; megakaryoblastic leukemia (translocation) 1 homolog; megakaryoblastic leukemia 1 met; megakaryoblastic leukemia 1 protein; Megakaryoblastic leukemia 1 protein homolog; Megakaryocytic acute leukemia protein; Megakaryocytic acute leukemia protein homolog; MELODY; mixed lineage leukemia; MKL (megakaryoblastic leukemia)/myocardin-like 1; MKL/myocardin-like protein 1; MKL1; MKL1-elongated derivative of yield protein; MKL1met; MRTFA; MRTF-A; myocardin related transcription factor A; myocardin-related transcription factor A |
| Gene Symbols | Mrtfa |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.