Learn More
MKL1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302202A

Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene interacts with the transcription factor myocardin, a key regulator of smooth muscle cell differentiation. The encoded protein is predomitly nuclear and may help transduce signals from the cytoskeleton to the nucleus. This gene is involved in a specific translocation event that creates a fusion of this gene and the RNA-binding motif protein-15 gene. This translocation has been associated with acute megakaryocytic leukemia. [provided by RefSeq].Specifications
MKL1 | |
Polyclonal | |
Unconjugated | |
MRTFA | |
megakaryoblastic leukemia translocation 1, BSAC, MAL, MRTF-A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57591 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q969V6 | |
MRTFA | |
Between 750 and 800 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.