Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mkl1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304625100UL
Description
Mkl1 Polyclonal specifically detects Mkl1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Mkl1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
57591 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
AMKL, KIAA1438, MALSAP and coiled-coil domain, megakaryoblastic leukemia (translocation) 1, Megakaryoblastic leukemia 1 protein, Megakaryocytic acute leukemia protein, MKL/myocardin-like protein 1, Myocardin-related transcription factor A, RNA-binding motif protein 15/megakaryoblastic leukemia-1 fusion protein | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human MKL1 (NP_065882.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction