Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MKL2 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP146208AF405
Description
MKL2 Polyclonal antibody specifically detects MKL2 in Human samples. It is validated for Western BlotSpecifications
MKL2 | |
Polyclonal | |
Western Blot | |
DKFZp686J1745, FLJ31823, FLJ45623, KIAA1243, megakaryoblastic leukemia 2, MKL/myocardin-like 2, MKL/myocardin-like protein 2, MRTFB, MRTF-B, myocardin-related transcription factor B, NPD001 | |
The immunogen for this product maps to a region between residue 900 and 950 of human Megakaryoblastic Leukemia 2 using the numbering given in entry NP_054767.3 (GeneID 57496). | |
0.1 mL | |
Cardiovascular Biology, Stem Cells | |
57496 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction