Learn More
Invitrogen™ MKL2 Monoclonal Antibody (CL1546)

Description
Immunogen sequence: QRPGPMELVE KNILPVDSSV KEAIIGVGKE DYPHTQGDFS FDEDSSDALS PDQPASQESQ GSAASPSEPK VSESPSPVTT NTPAQFASVS PTVPEFLKTP PTADQPPPRP AAPVLPTNTV SSAKPGPALV KQSHPKNP Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 54%. Binds to an epitope located within the peptide sequence FASVSPTVPEFLKTP as determined by overlapping synthetic peptides.
Specifications
Specifications
| Antigen | MKL2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | CL1546 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Mkl2 |
| Gene Accession No. | Q9ULH7 |
| Gene Alias | CTA-276F8.1; ENSMUSG00000075401; gene trap insertion site 4-1; Gt4-1; KIAA1243; Megakaryoblastic leukemia 2; megakaryoblastic leukemia 2 protein; mKIAA1243; MKL/myocardin like 2; MKL/myocardin-like 2; MKL/myocardin-like protein 2; MKL1/myocardin like 2; Mkl2; Mrtfb; MRTF-B; myocardin related transcription factor B; myocardin-related transcription factor B; myocardin-related transcription factor B; MKL/myocardin-like protein 2; NPD001; RGD1560833 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.