Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MKL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103370
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103370 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103370 Supplier Invitrogen™ Supplier No. PA5103370
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MKL2.

Serum response factor (SRF) is a transcription factor that binds the serum response element (SRE), a sequence that mediates the transient response of many cellular genes to growth stimulation. SRF regulates the transient response of several muscle genes in response to growth factors and recruits accessory myogenic factors to activate these muscle genes. SRF is required for the formation of vertebrate mesoderm leading to the origin of the cardiovascular system. Myocardin, in association with SRF in cardiac muscle cells, activates cardiac muscle promoters. Myocardin-related transcription factors A and B (MRTF-A and MRTF-B) interact with SRF and act as stimulators for its transcriptional activity. MRTF-B is crucial for skeletal myogenic differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MKL2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Mkl2
Gene Accession No. P59759, Q9ULH7
Gene Alias CTA-276F8.1; ENSMUSG00000075401; gene trap insertion site 4-1; Gt4-1; KIAA1243; Megakaryoblastic leukemia 2; megakaryoblastic leukemia 2 protein; mKIAA1243; MKL/myocardin like 2; MKL/myocardin-like 2; MKL/myocardin-like protein 2; MKL1/myocardin like 2; Mkl2; Mrtfb; MRTF-B; myocardin related transcription factor B; myocardin-related transcription factor B; myocardin-related transcription factor B; MKL/myocardin-like protein 2; NPD001; RGD1560833
Gene Symbols Mkl2
Host Species Rabbit
Immunogen A synthesized peptide derived from human MKL2(Accession Q9ULH7), corresponding to amino acid residues A605-S655.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 239719, 57496
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.