Learn More
MKL2 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302768A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Serum response factor (SRF) is a transcription factor that binds the serum response element (SRE), a sequence that mediates the transient response of many cellular genes to growth stimulation. SRF regulates the transient response of several muscle genes in response to growth factors and recruits accessory myogenic factors to activate these muscle genes. SRF is required for the formation of vertebrate mesoderm leading to the origin of the cardiovascular system. Myocardin, in association with SRF in cardiac muscle cells, activates cardiac muscle promoters. Myocardin-related transcription factors A and B (MRTF-A and MRTF-B) interact with SRF and act as stimulators for its transcriptional activity. MRTF-B is crucial for skeletal myogenic differentiation.Specifications
MKL2 | |
Polyclonal | |
Unconjugated | |
MKL2 | |
MKL/myocardin-like protein 2, Megakaryoblastic leukemia 2, Myocardin-related transcription factor B, MRTF-B, MKL2, KIAA1243, MRTFB | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
239719, 57496 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P59759, Q9ULH7 | |
MKL2 | |
Between 1038 and 1088 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.