Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKLP1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988981
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8981 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8981 Supplier Bioss Supplier No. BS9315R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a member of kinesin-like protein family. This family includes microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. This protein has been shown to cross-bridge antiparallel microtubules and drive microtubule movement in vitro. Alternate splicing of this gene results in two transcript variants encoding two different isoforms.

Specifications

Antigen MKLP1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KIF23
Gene Accession No. Q02241
Gene Alias 3110001D19Rik; BOS_10183; C87313; cb738; CHO1; KIF23; kif23 protein; kinesin family member 23; kinesin-like 5; kinesin-like 5 (mitotic kinesin-like protein 1); Kinesin-like protein 5; Kinesin-like protein KIF23; KNSL5; Mitotic kinesin-like protein 1; MKLP1; MKLP-1; wu:fi30e02; wu:fi39f08; zMKLP1
Gene Symbols KIF23
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MKLP1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9493
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.