Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKNK1, Mouse anti-Human, Clone: 24C2, Abnova™

Catalog No. 89998953 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-953 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-953 Supplier Abnova Corporation Supplier No. MAB16221
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen MKNK1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 24C2
Conjugate Unconjugated
Dilution ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:400) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In HEPES, 150 mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide).
Gene MAP kinase interacting serine/threonine kinase 1
Gene Alias MNK1
Gene Symbols MKNK1
Host Species Mouse
Immunogen Recombinant His fusion protein corresponding to amino acids 1-362 of human MKNK1 from E. coli.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8569
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.