Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MKP-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116976
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116976 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116976 Supplier Invitrogen™ Supplier No. PA5116976
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HepG2, BT-474, Mouse brain Immunogen sequence: MVMEVGTLDA GGLRALLGER AAQCLLLDCR SFFAFNAGHI AGSVNVRFST IVRRRAKGAM GLEHIVPNAE LRGRLLAGAY HAVVLLDERS AALDGAKRDG TLALAAGALC REARAAQVFF LKGGYEAFSA SCPELCSKQS TPMGLSLPLS TSVPDSAESG.

The expression of DUSP1 gene is induced in human skin fibroblasts by oxidative/heat stress and growth factors. It specifies a protein with structural features similar to members of the non-receptor-type protein-tyrosine phosphatase family, and which has significant amino-acid sequence similarity to a Tyr/Ser-protein phosphatase encoded by the late gene H1 of vaccinia virus. The bacterially expressed and purified DUSP1 protein has intrinsic phosphatase activity, and specifically inactivates mitogen-activated protein (MAP) kinase in vitro by the concomitant dephosphorylation of both its phosphothreonine and phosphotyrosine residues. Furthermore, it suppresses the activation of MAP kinase by oncogenic ras in extracts of Xenopus oocytes. Thus, DUSP1 may play an important role in the human cellular response to environmental stress as well as in the negative regulation of cellular proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MKP-1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Dusp1
Gene Accession No. P28562, P28563, Q64623
Gene Alias 3ch134; 3CH134/CL100 PTPase; CL 100; CL100; dual specificity phosphatase 1; dual specificity protein phosphatase 1; Dual specificity protein phosphatase hVH1; DUS1; DUSP1; EC 3.1.3.16; EC 3.1.3.48; erp; HVH1; MAP kinase phosphatase 1; MAP kinase phosphatase-1; mitogen-activated protein (MAP) kinase phosphatase-1; mitogen-activated protein kinase phosphatase 1; mitogen-activated protein kinase phosphatase-1; MKP1; mkp-1; oxidative stress-inducible protein tyrosine phosphatase; protein tyrosine phosphatase non-receptor type 16; protein tyrosine phosphatase, non-receptor type 16; protein-tyrosine phosphatase 3CH134; Protein-tyrosine phosphatase CL100; protein-tyrosine phosphatase ERP; Protein-tyrosine phosphatase non-receptor type 16; PTPN10; Ptpn16; serine/threonine specific protein phosphatase; U19515; VH1
Gene Symbols Dusp1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human DUSP1/MKP1 (NP_0044081).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114856, 1843, 19252
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.