Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MKP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598425
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598425 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598425 Supplier Invitrogen™ Supplier No. PA598425
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MKP2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

DUSP4 is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP) kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which are associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. DUSP4 inactivates ERK1, ERK2 and JNK, is expressed in a variety of tissues, and is localized in the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MKP2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene DUSP4
Gene Accession No. Q13115, Q8BFV3
Gene Alias 2700078F24Rik; AI844617; BB104621; dual specificity phosphatase 4; dual specificity protein phosphatase 4; Dual specificity protein phosphatase hVH2; DUSP 4; DUSP4; DUSP-4; E130306H24Rik; HVH2; MAP kinase phosphatase 2; mitogen-activated protein kinase phosphatase 2; MKP II; Mkp2; Mkp-2; MKPII; serine/threonine specific protein phosphatase; TYP; VH1 homologous phosphatase 2; VH2
Gene Symbols DUSP4
Host Species Rabbit
Immunogen Recombinant Human Dual specificity protein phosphatase 4 protein (109-198AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1846, 319520
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.