Learn More
Invitrogen™ MKP2 Polyclonal Antibody

Description
MKP2 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | MKP2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | DUSP4 |
| Gene Accession No. | Q13115, Q8BFV3 |
| Gene Alias | 2700078F24Rik; AI844617; BB104621; dual specificity phosphatase 4; dual specificity protein phosphatase 4; Dual specificity protein phosphatase hVH2; DUSP 4; DUSP4; DUSP-4; E130306H24Rik; HVH2; MAP kinase phosphatase 2; mitogen-activated protein kinase phosphatase 2; MKP II; Mkp2; Mkp-2; MKPII; serine/threonine specific protein phosphatase; TYP; VH1 homologous phosphatase 2; VH2 |
| Gene Symbols | DUSP4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.