Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MKP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582081
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582081 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582081 Supplier Invitrogen™ Supplier No. PA582081
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ECPHLCETSL AGRAGSSMAP VPGPVPVVGL GSLCLGSDCS DAESEADRDS MSCGLDSEGA TPPPVGLRAS FPVQILPNLY LGSARDSANL ESLAKLGIRY ILNVTPNLPN FFEKNGDFHY KQI Highest antigen sequence indentity to the following orthologs: Mouse - 82%, Rat - 80%.

Mitogen-activated protein (MAP) kinases are a large class of proteins involved in signal transduction pathways that are activated by a range of stimuli and mediate a number of physiological and pathological changes in the cell. Dual specificity phosphatases (DSPs) are a subclass of the protein tyrosine phosphatase (PTP) gene superfamily, which are selective for dephosphorylating critical phosphothreonine and phosphotyrosine residues within MAP kinases. DSP gene expression is induced by a host of growth factors and/or cellular stresses, thereby negatively regulating MAP kinase superfamily members including MAPK/ERK, SAPK/JNK and p38. The members of the dual-specificity phosphatase protein family include MKP-1/CL100 (3CH134), VHR, PAC1, MKP-2, hVH-3 (B23), hVH-5, MKP-3, MKP-X and MKP-4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MKP4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DUSP9
Gene Accession No. Q99956
Gene Alias dual specificity phosphatase 9; dual specificity protein phosphatase 9; dual-specificity MAP kinase phosphatase 4; Dusp4; DUSP9; MAP kinase phosphatase 4; mitogen-activated protein kinase phosphatase 4; MKP4; MKP-4; Mpk4; Pyst3; serine/threonine specific protein phosphatase
Gene Symbols DUSP9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DUSP9. Recombinant protein control fragment (Product #RP-92206).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1852
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.