Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MKRN1 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10055304FR
Description
MKRN1 Polyclonal antibody specifically detects MKRN1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
MKRN1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
E3 ubiquitin-protein ligase makorin-1, EC 6.3.2.-, FLJ21334, makorin ring finger protein 1, RNF61RING finger protein 61 | |
The immunogen recognized by this antibody maps to a region between residue 432 and the C-terminus (residue 482) of human makorin, ring finger protein, 1 using the numbering given in entry NP_038474.1 (GeneID 23608). | |
0.1 mL | |
Zinc Finger | |
23608 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction