Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MKRN1 Antibody (OTI2C8), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272718MFV500
Description
MKRN1 Monoclonal antibody specifically detects MKRN1 in Human, Mouse samples. It is validated for Flow CytometrySpecifications
MKRN1 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry | |
OTI2C8 | |
Flow Cytometry | |
E3 ubiquitin-protein ligase makorin-1, EC 6.3.2.-, FLJ21334, makorin ring finger protein 1, RNF61RING finger protein 61 | |
Full length human recomnbinant protein of human MKRN1(NP_038474) produced in HEK293T cell. | |
0.1 mL | |
Zinc Finger | |
23608 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction