Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MKRN1 Antibody (OTI3F9), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272716UV
Description
MKRN1 Monoclonal specifically detects MKRN1 in Human, Mouse samples. It is validated for Western Blot.Specifications
MKRN1 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MKRN1 | |
Full length recombinant protein of human MKRN1 (NP_038474) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG2b |
Western Blot | |
OTI3F9 | |
Western Blot | |
E3 ubiquitin-protein ligase makorin-1, EC 6.3.2.-, FLJ21334, makorin ring finger protein 1, RNF61RING finger protein 61 | |
Mouse | |
Affinity Purified | |
Zinc Finger | |
23608.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction