Learn More
Invitrogen™ MKRN1 Monoclonal Antibody (4E9)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA521521
Catalog No. PIMA521521
Only null left
Add to Cart
Description
Description
Peptide Sequence: MSNKACRYFD EGRGSCPFGG NCFYKHAYPD GRREEPQRQK VGTSSRYRAQ RRNHFWELIE ERENSNPFDN DEEEVV.
The Makorin ring finger protein-1 gene (MKRN1) is a highly transcribed, intron-containing source for a family of intronless mammalian genes encoding a novel class of zinc finger proteins. Phylogenetic analyses indicate that the MKRN1 gene is the ancestral founder of this gene family.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MKRN1
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q9UHC7
MKRN1
MKRN1 (NP_038474, 365 a.a. ~ 440 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2b κ
ELISA, Western Blot
4E9
Unconjugated
MKRN1
E3 ubiquitin-protein ligase makorin-1; makorin ring finger protein 1; MKRN1; RING finger protein 61; RING-type E3 ubiquitin transferase makorin-1; RNF61
Mouse
Affinity chromatography
RUO
23608
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.