Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MKRN1 Monoclonal Antibody (OTI3F9), TrueMAB™, OriGene
SDP

Catalog No. 502540884
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0884 30 μL
50-167-4368 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0884 Supplier OriGene Supplier No. TA503486S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MKRN1 Monoclonal antibody specifically detects MKRN1 in Human samples. It is validated for Western Blot

Specifications

Antigen MKRN1
Applications Western Blot
Classification Monoclonal
Clone OTI3F9
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CYP2J2
Gene Accession No. P51589
Gene Alias E3 ubiquitin-protein ligase makorin-1; makorin ring finger protein 1; MKRN1; RING finger protein 61; RING-type E3 ubiquitin transferase makorin-1; RNF61
Host Species Mouse
Immunogen Full length human recombinant protein of human CYP2J2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1573
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.