Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MKRN1 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142736
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize both reported isoforms (NP_038474.2; NP_001138597.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
The Makorin ring finger protein-1 gene (MKRN1) is a highly transcribed, intron-containing source for a family of intronless mammalian genes encoding a novel class of zinc finger proteins. Phylogenetic analyses indicate that the MKRN1 gene is the ancestral founder of this gene family.
Specifications
MKRN1 | |
Polyclonal | |
Unconjugated | |
MKRN1 | |
E3 ubiquitin-protein ligase makorin-1; makorin ring finger protein 1; MKRN1; RING finger protein 61; RING-type E3 ubiquitin transferase makorin-1; RNF61 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
23608, 54484 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9QXP6, Q9UHC7 | |
MKRN1 | |
Peptide with sequence C-RYEHSKPLKQEEAT. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction