Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MKRN2 Monoclonal Antibody (1556CT631.230.55.49)
GREENER_CHOICE

Catalog No. PIMA524741
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524741 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524741 Supplier Invitrogen™ Supplier No. MA524741
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MKRN2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

Members of the makorin family, including MKRN2, have a characteristic zinc finger composition that suggests that they are ribonucleoproteins (Gray et al., 2001).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MKRN2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1556CT631.230.55.49
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MKRN2
Gene Accession No. Q9H000
Gene Alias 2610002L04Rik; C81377; HSPC070; makorin ring finger protein 2; Makorin RING zinc-finger protein 2; makorin, ring finger protein, 2; Mkrn2; Probable E3 ubiquitin-protein ligase makorin-2; RING finger protein 62; RING-type E3 ubiquitin transferase makorin-2; RNF62
Gene Symbols MKRN2
Host Species Mouse
Immunogen human MKRN2 recombinant protein.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23609
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.