Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MKRN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558004
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558004 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558004 Supplier Invitrogen™ Supplier No. PA558004
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NSHEPGKREK RTLVLRDRNL SGMAERKTQP SMVSNPGSCS DPQPSPEMKP HSYLDAIRSG LDDVEASSSY SNEQQ Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 79%.

Members of the makorin family, including MKRN2, have a characteristic zinc finger composition that suggests that they are ribonucleoproteins (Gray et al., 2001).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MKRN2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MKRN2
Gene Accession No. Q9ERV1, Q9H000
Gene Alias 2610002L04Rik; C81377; HSPC070; makorin ring finger protein 2; Makorin RING zinc-finger protein 2; makorin, ring finger protein, 2; Mkrn2; Probable E3 ubiquitin-protein ligase makorin-2; RING finger protein 62; RING-type E3 ubiquitin transferase makorin-2; RNF62
Gene Symbols MKRN2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MKRN2. Recombinant protein control fragment (Product #RP-96289).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23609, 297525, 67027
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.