Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MKRN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558005
Description
Immunogen sequence: ACKYFEQGKG TCPFGSKCLY RHAYPDGRLA EPEKPRKQLS SQGTVRFFNS VRLWDFIENR ESRHVPNNED VDMTELGDLF MHLSGVESS Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
Members of the makorin family, including MKRN2, have a characteristic zinc finger composition that suggests that they are ribonucleoproteins (Gray et al., 2001).
Specifications
MKRN2 | |
Polyclonal | |
Unconjugated | |
MKRN2 | |
2610002L04Rik; C81377; HSPC070; makorin ring finger protein 2; Makorin RING zinc-finger protein 2; makorin, ring finger protein, 2; Mkrn2; Probable E3 ubiquitin-protein ligase makorin-2; RING finger protein 62; RING-type E3 ubiquitin transferase makorin-2; RNF62 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23609 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H000 | |
MKRN2 | |
Recombinant protein corresponding to Human MKRN2. Recombinant protein control fragment (Product #RP-96290). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction