Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MKRN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530482
Description
Recommended positive controls: HeLa. Predicted reactivity: Mouse (82%), Rat (82%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Members of the makorin family, including MKRN2, have a characteristic zinc finger composition that suggests that they are ribonucleoproteins (Gray et al., 2001).
Specifications
| MKRN2 | |
| Polyclonal | |
| Unconjugated | |
| MKRN2 | |
| 2610002L04Rik; C81377; HSPC070; makorin ring finger protein 2; Makorin RING zinc-finger protein 2; makorin, ring finger protein, 2; Mkrn2; Probable E3 ubiquitin-protein ligase makorin-2; RING finger protein 62; RING-type E3 ubiquitin transferase makorin-2; RNF62 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23609 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.74 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9H000 | |
| MKRN2 | |
| Recombinant protein encompassing a sequence within the center region of human MKRN2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction