Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MKRN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585989
Description
MKRN2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Members of the makorin family, including MKRN2, have a characteristic zinc finger composition that suggests that they are ribonucleoproteins (Gray et al., 2001).
Specifications
| MKRN2 | |
| Polyclonal | |
| Unconjugated | |
| MKRN2 | |
| 2610002L04Rik; C81377; HSPC070; makorin ring finger protein 2; Makorin RING zinc-finger protein 2; makorin, ring finger protein, 2; Mkrn2; Probable E3 ubiquitin-protein ligase makorin-2; RING finger protein 62; RING-type E3 ubiquitin transferase makorin-2; RNF62 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23609, 67027 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9ERV1, Q9H000 | |
| MKRN2 | |
| Synthetic peptide corresponding to Human MRKN2 aa 281-330. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction