Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MKRN2OS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564247
Description
Immunogen sequence: YDGRSDLHVG ITNTNGVVYN YSAHGVQRDG EGWEESISIP LLQPNMYGMM EQWDKYLEDF STSGAWLPHR YEDNH Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 73%.

Specifications
MKRN2OS | |
Polyclonal | |
Unconjugated | |
MKRN2OS | |
C3orf83; MKRN2 antisense gene protein 1; MKRN2 antisense RNA 1; MKRN2 antisense RNA 1 (non-protein coding); MKRN2 opposite strand; MKRN2 opposite strand protein; MKRN2-AS1; MKRN2OS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100129480 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
H3BPM6 | |
MKRN2OS | |
Recombinant protein corresponding to Human MKRN2OS. Recombinant protein control fragment (Product #RP-104069). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction