Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MKRN3 Monoclonal Antibody (2E10)
GREENER_CHOICE

Catalog No. PIMA520268
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA520268 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA520268 Supplier Invitrogen™ Supplier No. MA520268
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: SFSAYWHQLV EPVRMGEGNM LYKSIKKELV VLRLASLLFK RFLSLRDELP FSEDQWDLLH YELEEYFNLI L.

The protein encoded by this gene contains a RING (C3HC4) zinc finger motif and several C3H zinc finger motifs. This gene is intronless and imprinted, with expression only from the paternal allele. Disruption of the imprinting at this locus may contribute to Prader-Willi syndrome. An antisense RNA of unknown function has been found overlapping this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MKRN3
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 2E10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene MKRN3
Gene Accession No. Q13064
Gene Alias CPPB2; D15S9; D7H15S9-1; makorin ring finger protein 3; makorin, ring finger protein, 3; Mkrn3; Probable E3 ubiquitin-protein ligase makorin-3; RING finger protein 63; RING-type E3 ubiquitin transferase makorin-3; RNF63; Zfp127; Zinc finger protein 127; ZNF127
Gene Symbols MKRN3
Host Species Mouse
Immunogen MKRN3 (NP_005655, 437 a.a. ~ 507 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7681
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.