Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MKS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120892
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120892 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120892 Supplier Invitrogen™ Supplier No. PA5120892
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: MCF7, BT-474, A-549, SW480, Mouse testis, Mouse ovary, Mouse small intestine, Rat brain, Rat liver. The target is usually found in the following locations: Cytoplasm, centrosome, cilium basal body, cytoskeleton, microtubule organizing center. Immunogen sequence: EVPSFLVERM ANVRRRRQDR RGMEGGILKS RIVTWEPSEE FVRNNHVINT PLQTMHIMAD LGPYKKLGYK KYEHVLCTLK VDSNGVITVK PDFTGLKGPY RIETEGEKQE LWKYTIDNVS PHAQPEEEER ERRVFKDLYG RHKEYLSSLV G.

The protein encoded by this gene localizes to the basal body and is required for formation of the primary cilium in ciliated epithelial cells. Mutations in this gene result in Meckel syndrome type 1 and in Bardet-Biedl syndrome type 13. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MKS1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MKS1
Gene Accession No. Q499Q5, Q5SW45, Q9NXB0
Gene Alias AK190930; B8d3; BBS13; LOW QUALITY PROTEIN: Meckel syndrome type 1 protein; Meckel syndrome type 1 protein; Meckel syndrome type 1 protein homolog; Meckel syndrome, type 1; MES; MKS; MKS transition zone complex subunit 1; Mks1; POC12; POC12 centriolar protein homolog
Gene Symbols MKS1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 150-300 of human MKS1 (NP_060247.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287612, 380718, 54903
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.