Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MKX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540565
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540565 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540565 Supplier Invitrogen™ Supplier No. PA540565
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: IKSENSVIKA GVRPESRASE DYVAPPKYKS SLLNRYLNDS LRHVMATNTT Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 92%; Rabbit: 93%; Rat: 92%; Zebrafish: 85%.

MKX is a member of an Iroquois (IRX) family-related class of 'three-amino acid loop extension' (TALE) atypical homeobox proteins characterized by 3 additional amino acids in the loop region between helix I and helix II of the homeodomain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MKX
Applications Immunohistochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MKX
Gene Accession No. Q8IYA7
Gene Alias 9430023B20Rik; C10orf48; homeobox protein mohawk; IFRX; Iroquois family related homeodomain protein; iroquois homeobox protein-like 1; Irxl1; MKX; mohawk homeobox; RGD1305652
Gene Symbols MKX
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human MKX.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283078
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.