Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 00253764 hydrochloride
GREENER_CHOICE

Catalog No. 485410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-541-0 10 mg
48-545-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-541-0 Supplier Tocris Bioscience™ Supplier No. 4854/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Melanocortin MC4 receptor antagonist; brain penetrant

ML 00253764 hydrochloride is a non-peptide, melanocortin MC4 receptor antagonist (IC50 values are 320, 810 and 2120 nM for MC4, MC3 and MC5 receptors respectively). Decreases cAMP accumulation in HEK-293 cells expressing the MC4 receptor. Increases food intake and reduces loss of lean body mass in tumor bearing mice. Brain penetrant.

Specifications

Quantity 10 mg
Product Type ML 00253764 hydrochloride
Purity >98%
Molecular Weight (g/mol) 413.71
Inhibitors ML 00253764 hydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.