Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 141
GREENER_CHOICE

Catalog No. 426610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-661-0 10 mg
42-665-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-661-0 Supplier Tocris Bioscience™ Supplier No. 4266/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective Cdc42 Rho family inhibitor

Allosteric inhibitor of Cdc42 GTPase (EC50 = 2.1 μM). Selective for Cdc42 over other members of the Rho GTPase family including Rac1, Rab2 and Rab7. Inhibits ovarian cancer cell migration in vitro.

Chemical Identifiers

CAS 71203-35-5
Molecular Formula C22H21N3O3S

Specifications

Quantity 10 mg
Formula Weight 407.49
Purity >99%
Molecular Weight (g/mol) 407.488
Chemical Name or Material Rho GTPase Inhibitors
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.