Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ML 162
GREENER_CHOICESDP

Catalog No. 782110 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
78-211-0 10 mg
RD10195097 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 78-211-0 Supplier R&D Systems™ Supplier No. 7821/10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibits glutathione peroxidase (GPX4); induces ferroptosis

ML 162 is a covalent glutathione peroxidase 4 (GPX4) inhibitor. It induces ferroptosis in vivo. It is selectively lethal to mutant RAS oncogene-expressing cell lines (IC50 = 25 and 578 nM for HRASG12V-expressing and wild-type BJ fibroblasts, respectively).

Specifications

Chemical Name or Material 2-[(Chloroacetyl)(3-chloro-4-methoxyphenyl)amino]-N-(2-phenylethyl)-2-thien-2-ylacetamide
CAS 1035072-16-2
Quantity 10 mg
Product Line Tocris
Target Glutathione Peroxidase 4/GPX4
Molecular Formula C23H22Cl2N2O3S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.