Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ML 193
GREENER_CHOICESDP

Catalog No. 486010 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
486010 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 486010 Supplier R&D Systems™ Supplier No. 4860/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, selective GPR55 antagonist

ML 193 is a selective GPR55 antagonist (IC50 = 221 nM). Exhibits >27-, >145- and >145-fold selectivity for GPR55 over CB1, GPR35 and CB2, respectively. Inhibits ERK signaling in vitro.

Specifications

Chemical Name or Material ML 193
CAS 713121-80-3
Synonym N-[4-[[(3,4-Dimethyl-5-isoxazolyl)amino]sulfonyl]phenyl]-6,8-dimethyl-2-(2-pyridinyl)-4-quinolinecarboxamide
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Product Line Tocris
Molecular Formula C28H25N5O4S
Formula Weight 527.59
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.