Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (±)-ML 209
GREENER_CHOICE

Catalog No. 598710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-871-0 10 mg
59-875-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 59-871-0 Supplier Tocris Bioscience™ Supplier No. 5987/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

RORγt inverse agonist; suppresses Th17 cell differentiation

(±)-ML 209 is a RORγt inverse agonist (IC50 = 460 nM). Suppresses Th17 cell differentiation and IL17A expression in vitro. Does not suppress transcription of RORα. Minimal activity displayed on ERα, LXRα and thyroid hormone receptors (IC50 values >4.4 μM).

Specifications

CAS 1334526-14-5
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Target Nuclear Receptor Antagonists
Molecular Formula C25H31NO6
Formula Weight 441g/mol
Color White
Product Type Inhibitor

For Laboratory Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.