Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ML 210
GREENER_CHOICESDP

Catalog No. 642910 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-291-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 64-291-0 Supplier R&D Systems™ Supplier No. 6429/10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selectively kills mutant HRAS-expressing cells

ML 210 is a glutathione peroxidase (GPX4) inhibitor. Kills mutant RAS-expressing cell lines (IC50 values are 71 and 272 nM for HRAS G12v mutant expressing cell lines BJeLR and BJeH-LT, respectively). Exhibits 4-fold selectivity for HRAS mutant-expressing cell lines. Induces ferroptosis in tumor initiation "persister" cells and drug-resistant tumor cells.

Specifications

Chemical Name or Material [4-[Bis(4-chlorophenyl)methyl]-1-piperazinyl](5-methyl-4-nitro-3-isoxazolyl)methanone
CAS 1360705-96-9
Quantity 10 mg
Product Line Tocris
Molecular Formula C22H20Cl2N4O4
Formula Weight 475.32
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.