Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 213
GREENER_CHOICE

Catalog No. 451910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-191-0 10 mg
45-195-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-191-0 Supplier Tocris Bioscience™ Supplier No. 4519/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

KV7.2 and KV7.4 channel opener

Selective KV7.2 (KCNQ2) and KV7.4 (KCNQ4) channel opener (EC50 values are 230 and 510 nM for KV7.2 and KV7.4 respectively). Displays > 80-fold selectivity against KV7.1, KV7.3 and KV7.5 in a thallium-based fluorescence assay. Hyperpolarizes resting membrane potential in an in vitro spinal cord preparation.

Chemical Identifiers

CAS 489402-47-3
Molecular Formula C17H23NO

Specifications

Quantity 10 mg
Formula Weight 257.37
Purity >99%
Molecular Weight (g/mol) 257.377
Chemical Name or Material ML 213
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.