Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ML 240
GREENER_CHOICESDP

Catalog No. 515350 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-535-0 50 mg
51-531-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 51-535-0 Supplier R&D Systems™ Supplier No. 5153/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. ATP-competitive inhibitor of p97 ATPase

ML 240 is an ATP-competitive inhibitor of p97 ATPase (VCP, IC50 = 110 nM). Exhibits antiproliferative activity in NCI-60 cancer cell lines and rapidly induces executioner caspases 3 and 7 in multiple colon cancer cells. Promotes accumulation of LC3-II and impairs autophagosome maturation. Also impairs the endoplasmic-reticulum-associated degradation (ERAD) pathway.

Specifications

Quantity 50 mg
Product Line Tocris
Product Type ML 240
Purity >99%
Molecular Weight (g/mol) 396.44
Inhibitors ML 240

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.