Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 277
GREENER_CHOICE

Catalog No. 477710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-771-0 10 mg
47-775-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-771-0 Supplier Tocris Bioscience™ Supplier No. 4777/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective Kv7.1 (KCNQ1) potassium channel activator; augments IKs current

ML 277 is a selective Kv7.1 (KCNQ1) potassium channel activator (EC50 = 260 nM). Exhibits >100-fold selectivity versus KCNQ2, KCNQ4 and hERG potassium channels. Augments IKs current of cultured human cardiomyocytes and shortens action potential duration.

Specifications

Quantity 10 mg
Product Type ML 277
Purity >98%
Molecular Weight (g/mol) 471.59
Inhibitors ML 277

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.