Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 281
GREENER_CHOICE

Catalog No. 488010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-801-0 10 mg
48-805-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-801-0 Supplier Tocris Bioscience™ Supplier No. 4880/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

STK33 inhibitor

Inhibitor of the serine/threonine kinase STK33 (IC50 = 14 nM). Exhibits >700-fold selectivity for STK33 over the structurally related protein kinase A (PKA); also exhibits 550-fold selectivity over Aurora kinase B.

Chemical Identifiers

CAS 1404437-62-2
Molecular Formula C22H19N3O2S

Specifications

Quantity 10 mg
Formula Weight 389.47
Purity >98%
Molecular Weight (g/mol) 389.473
Chemical Name or Material ML 281
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.