Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ML 281
GREENER_CHOICESDP

Catalog No. 488050 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-805-0 50 mg
48-801-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-805-0 Supplier R&D Systems™ Supplier No. 4880/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. STK33 inhibitor

Inhibitor of the serine/threonine kinase STK33 (IC50 = 14 nM). Exhibits >700-fold selectivity for STK33 over the structurally related protein kinase A (PKA); also exhibits 550-fold selectivity over Aurora kinase B.

Chemical Identifiers

CAS 1404437-62-2
Molecular Formula C22H19N3O2S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 389.47
Purity >98%
Molecular Weight (g/mol) 389.473
Chemical Name or Material ML 281
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.