Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 297
GREENER_CHOICE

Catalog No. 538010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-801-0 10 mg
53-805-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 53-801-0 Supplier Tocris Bioscience™ Supplier No. 5380/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective Kir3.1/3.2 (GIRK1/2) channel activator

ML 297 is a selective Kir3.1/3.2 (GIRK1/2) channel activator (IC50 values are 160, 887 and 914 nM for GIRK1/2, GIRK1/4 and GIRK1/3 respectively). Exhibits no effect on GIRK2, GIRK2/3, Kir2.1 and Kv7.4 channels, and has minimal effect on a panel of other ion channels, receptors and transporters. Prevents PTZ -induced epileptic seizures in mice. Brain penetrant.

Specifications

Quantity 10 mg
Product Type ML 297
Purity >99%
Molecular Weight (g/mol) 328.32
Inhibitors ML 297

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.