Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 335
GREENER_CHOICE

Catalog No. 688710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-871-0 10 mg
68-875-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 68-871-0 Supplier Tocris Bioscience™ Supplier No. 6887/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective K2P2.1 (TREK-1) and K2P10.1 (TREK-2) activator

ML 335 is a selective K2P2.1 (TREK-1; KCNK2) and K2P10.1 (TREK-2; KCNK10) activator (EC50 values of 14.3 μM and 5.2 μM, respectively); directly stimulates the C-type gate via the K2P modulator pocket. Does not activate K2P4.1 (TRAAK).

Specifications

Quantity 10 mg
Molecular Formula C15H14Cl2N2O3S
Formula Weight 373.25
Product Type Activator
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.