Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 337
GREENER_CHOICESDP

Catalog No. 498250R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-825-0R 50 mg
49-821-0R 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 49-825-0R Supplier Tocris Bioscience™ Supplier No. 4982/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective negative allosteric modulator of mGlu3

ML 337 is a selective negative allosteric modulator of mGlu3 (IC50 = 593 nM). Displays no activity at mGlu1, mGlu2 or mGlu4-8 at concentrations up to 30 μM. Brain penetrant.

Specifications

Quantity 50 mg
Product Type ML 337
Purity >98%
Molecular Weight (g/mol) 353.39
Inhibitors ML 337

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.