Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 382
GREENER_CHOICESDP

Catalog No. 573850 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-385-0 50 mg
57-381-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 57-385-0 Supplier Tocris Bioscience™ Supplier No. 5738/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

MrgX1 (SNSR4) positive allosteric modulator

ML 382 is a selective MRGPRX1 positive allosteric modulator (EC50 = 190 nM). Exhibits selectivity for MrgX1 over MrgX2, MrgC11 and a panel of 68 other GPCRs, ion channels and transporters. Enhances BAM (8-22) potency at MRGPRX1 by 7-fold.

Specifications

Chemical Name or Material Mas-Related G Protein Coupled Receptor Modulators
CAS 1646499-97-9
Quantity 50 mg
Target Sensory Neuron-Specific Receptor Agonists
Molecular Formula C18H20N2O4S
Formula Weight 360.43
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.