Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLANA Mouse anti-Human, Clone: 4F10D6, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | MLANA |
---|---|
Clone | 4F10D6 |
Concentration | 2 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6435
![]() ![]() |
Proteintech Group Inc
603481IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
MART1 is involved in melanosome biogenesis by ensuring the stability of GPR143. Plays a vital role in the expression, stability, trafficking, and processing of melanocyte protein PMEL, which is critical to the formation of stage II melanosomes.Specifications
MLANA | |
2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Antigen LB39 AA, Antigen SK29 AA, MART 1, MART1, melan A, MLANA, Protein Melan A | |
MLANA | |
IgG2a | |
Protein A | |
Antibody |
4F10D6 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
Q16655 | |
2315 | |
MLANA Fusion Protein Ag13387 | |
Primary | |
-20°C | |
MLANA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title