Learn More
MLANA Mouse anti-Human, Clone: OTI3A2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF506029

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-Ais a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Seven other melanoma associated antigens recognized by autologous cytotoxic T cells include MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1. Subcellular fractionation shows that MART-1 is present in melanosomes and endoplasmic reticulum.Specifications
MLANA | |
Monoclonal | |
Unconjugated | |
MLANA | |
MART-1, MART1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
2315 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI3A2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q16655 | |
MLANA | |
Full length human recombit protein of human MLANA produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.