Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP180073
Description
MLC1 Polyclonal specifically detects MLC1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MLC1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
LVM, megalencephalic leukoencephalopathy with subcortical cysts 1, WKL1 | |
Rabbit | |
41 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Guinea pig: 92%; Mouse: 92%; Rat: 92%; Canine: 85%; Equine: 85%; Rabbit: 83%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
NP_055981 | |
MLC1 | |
Synthetic peptide directed towards the middle region of human MLC1. Peptide sequence SDSANILDEVPFPARVLKSYSVVEVIAGISAVLGGIIALNVDDSVSGPHL. | |
Affinity purified | |
RUO | |
23209 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction