Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541042
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541042 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541042 Supplier Invitrogen™ Supplier No. PA541042
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SDSANILDEV PFPARVLKSY SVVEVIAGIS AVLGGIIALN VDDSVSGPHL Sequence homology: Cow: 91%; Dog: 86%; Guinea Pig: 92%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 83%; Rat: 93%.

MLC1 may be a transporter. It may act as a non-selective neuronal cation channel.The function of this gene product is unknown; however, homology to other proteins suggests that it may be an integral membrane transporter. Mutations in this gene have been associated with megalencephalic leukoencephalopathy with subcortical cysts, an autosomal recessive neurological disorder. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLC1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MLC1
Gene Accession No. Q15049
Gene Alias AW048630; BB074274; KIAA0027; Kiaa0027-hp; LVM; megalencephalic leukoencephalopathy with subcortical cysts 1; megalencephalic leukoencephalopathy with subcortical cysts 1 gene product; megalencephalic leukoencephalopathy with subcortical cysts 1 homolog; megalencephalic leukoencephalopathy with subcortical cysts 1 homolog (human); membrane protein MLC1; mKIAA0027; MLC; MLC1; modulator of VRAC current 1; VL; WKL1
Gene Symbols MLC1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human MLC1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23209, 315215
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.