Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLC1SA (MYL6B) Monoclonal Antibody (OTI5B11), TrueMAB™, OriGene
SDP

Catalog No. 502543954
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3954 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3954 Supplier OriGene Supplier No. CF811276
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MLC1SA (MYL6B) Monoclonal antibody specifically detects MLC1SA (MYL6B) in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen MLC1SA (MYL6B)
Applications Western Blot
Classification Monoclonal
Clone OTI5B11
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene SEPT3
Gene Accession No. Q9UH03, Q9Z1S5
Gene Alias 5730437E04Rik; BC037527; MLC1SA; MYL6B; myosin alkali light chain 1 slow a; myosin light chain 1 slow a; myosin light chain 1 slow-twitch muscle A isoform; myosin light chain 1, slow-twitch muscle A isoform; myosin light chain 6B; myosin, light chain 6B, alkali, smooth muscle and non-muscle; myosin, light polypeptide 6B; myosin, light polypeptide 6B, alkali, smooth muscle and non-muscle; sm; smooth muscle and non-muscle myosin alkali light chain 6B; smooth muscle and nonmuscle myosin light chain alkali 6B
Host Species Mouse
Immunogen Full length human recombinant protein of human SEPT3 produced in HEK293T cell.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24050, 55964
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.